Learn More
Cayman Chemical Lentztrehalose B, Purity: ≥80%, CAS: 1808096-67-4, 5 mg
SDP
Catalog No. 502916507
Encompass_Preferred
A disaccharide microbial metabolite with diverse biological activities; has antioxidant activity in an ORAC assay at 100 µM; inhibits the activity of porcine kidney trehalase at 10 mM; induces autophagy in MeWo and OVK18 cells at 100 mM
Catalog No. 50-291-6507 Supplier Cayman Chemical Supplier No. 377335 MG
A disaccharide microbial metabolite with diverse biological activities; has antioxidant activity in an ORAC assay at 100 µM; inhibits the activity of porcine kidney trehalase at 10 mM; induces autophagy in MeWo and OVK18 cells at 100 mM