Learn More
Cayman Chemical Lentztrehalose C, Purity: ≥75%, CAS: 1808096-68-5, 10 mg
SDP
Catalog No. 502916774
Encompass_Preferred
A disaccharidemicrobial metabolite with autophagy-inducing activity; induces autophagy in MeWo and OVK18 cells at 100 mM
Catalog No. 50-291-6774 Supplier Cayman Chemical Supplier No. 3773410 MG
A disaccharidemicrobial metabolite with autophagy-inducing activity; induces autophagy in MeWo and OVK18 cells at 100 mM