Learn More
Medchemexpress LLC HY-10981 10mg Medchemexpress, Lenvatinib CAS:417716-92-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1098110MG
Encompass_Preferred
Medchemexpress, HY-10981 10mg Lenvatinib CAS:417716-92-8 Lenvatinib (E7080) is an oral, multi-targeted tyrosine kinase inhibitor that inhibits VEGFR1-3, FGFR1-4, PDGFR, KIT, and RET, shows potent antitumor activities[2]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-8799