Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Lenvatinib-15N,d4 | 00-00-0 | 98.0% | 431.87 g/mol | C21H15D4ClN315NO4 | 5 MG
SDP

Catalog No. 5000264905
Encompass_Preferred

Lenvatinib-15N,d4 is an isotopically labeled analogue of lenvatinib intended for use as an internal standard or tracer in analytical and pharmacokinetic studies. The compound contains 15N and four deuterium labels to facilitate sensitive and specific LC-MS/MS quantification while limiting isotopic overlap.

  • Isotopically labeled with 15N and four deuteriums for use as an internal standard.
  • Formulated for analytical and pharmacokinetic LC-MS/MS applications.
  • High purity (98.01%) suitable for quantitative studies.
  • Powder form with recommended cold storage for stability.
  • Molecular weight 431.87 g/mol and molecular formula C21H15D4ClN315NO4.

Catalog No. 50-002-64905 Supplier Medchemexpress LLC Supplier No. HY10981S25MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Lenvatinib-15N,d4 is an isotopically labeled analogue of lenvatinib intended for use as an internal standard or tracer in analytical and pharmacokinetic studies. The compound contains 15N and four deuterium labels to facilitate sensitive and specific LC-MS/MS quantification while limiting isotopic overlap.

  • Isotopically labeled with 15N and four deuteriums for use as an internal standard.
  • Formulated for analytical and pharmacokinetic LC-MS/MS applications.
  • High purity (98.01%) suitable for quantitative studies.
  • Powder form with recommended cold storage for stability.
  • Molecular weight 431.87 g/mol and molecular formula C21H15D4ClN315NO4.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.