Learn More
Medchemexpress LLC Lenvatinib | 417716-92-8 | 99.8% | 1 G
SDP
Catalog No. 5000344960
Encompass_Preferred
Lenvatinib (E7080) is an oral, multi-targeted tyrosine kinase inhibitor. It inhibits VEGFR1-3, FGFR1-4, PDGFR, KIT, and RET, demonstrating potent antitumor activities. This compound shows antiproliferative activity and growth inhibition in various cell lines.
  • Inhibits VEGFR2, VEGFR3, and VEGFR1
  • Inhibits PDGFRα, PDGFRβ, and FGFR1
  • Inhibits KIT and RET
  • Demonstrates antiproliferative activity
  • Available as a solid form
Catalog No. 50-003-44960 Supplier Medchemexpress LLC Supplier No. HY10981200MG
Lenvatinib (E7080) is an oral, multi-targeted tyrosine kinase inhibitor. It inhibits VEGFR1-3, FGFR1-4, PDGFR, KIT, and RET, demonstrating potent antitumor activities. This compound shows antiproliferative activity and growth inhibition in various cell lines.
  • Inhibits VEGFR2, VEGFR3, and VEGFR1
  • Inhibits PDGFRα, PDGFRβ, and FGFR1
  • Inhibits KIT and RET
  • Demonstrates antiproliferative activity
  • Available as a solid form