Learn More
Cayman Chemical Lenvatinib-d4, Purity: ≥99% deuterated forms (d1-d4), CAS: 2264050-65-7, 1 mg
SDP
Catalog No. 502940353
Encompass_Preferred
An internal standard for the quantification of lenvatinib by GC- or LC-MS
Catalog No. 50-294-0353 Supplier Cayman Chemical Supplier No. 286151 MG
An internal standard for the quantification of lenvatinib by GC- or LC-MS