Learn More
Medchemexpress LLC Lenvatinib-d4 (E7080-d4) | 2264050-65-7 | 99.6% | 430.88 | 50 MG
SDP
Catalog No. 5000344929
Encompass_Preferred
Lenvatinib-d4 is the deuterium-labeled Lenvatinib. Lenvatinib (E7080) is an oral, multi-targeted tyrosine kinase inhibitor that inhibits VEGFR1-3, FGFR1-4, PDGFR, KIT, and RET, and shows potent antitumor activities.
  • Used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-003-44929 Supplier Medchemexpress LLC Supplier No. HY10981S1MG
Lenvatinib-d4 is the deuterium-labeled Lenvatinib. Lenvatinib (E7080) is an oral, multi-targeted tyrosine kinase inhibitor that inhibits VEGFR1-3, FGFR1-4, PDGFR, KIT, and RET, and shows potent antitumor activities.
  • Used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.