Learn More
Apexbio Technology LLC Lenvatinib (E7080), 5mg. Cas: 417716-92-8 MFCD: NA. VEGFR inhibitor
SDP
Catalog No. 501904735
Encompass_Preferred
Lenvatinib (E7080) is an inhibitor of multiple RTK with IC50 values of 22nM, 4nM, 5.2nM, 39nM and 35nM on VEGFR-1, VEGFR-2, VEGFR-3, PDGFR and RET, respectively [1]. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4735 Supplier Apexbio Technology LLC Supplier No. A21745
Lenvatinib (E7080) is an inhibitor of multiple RTK with IC50 values of 22nM, 4nM, 5.2nM, 39nM and 35nM on VEGFR-1, VEGFR-2, VEGFR-3, PDGFR and RET, respectively [1]. Other sizes are available. Please inqury us for quote.