Learn More
TARGETMOL CHEMICALS INC Lenvatinib mesylate 100MG
SDP
Catalog No. 502878120
Encompass_Preferred
Also available in 10 mg, 25 mg, 50 mg, 200 mg, 500 mg and bulk. Please contact Fisher for quotes. Lenvatinib mesylate (E7080 (mesylate)) is an oral and multi-targeted inhibitor of VEGFR1-3, FGFR1-4, PDGFR, KIT, and RET, with potent antitumor activities. Purity 99.03%
Catalog No. 50-287-8120 Supplier TARGETMOL CHEMICALS INC Supplier No. T8541100MG
Also available in 10 mg, 25 mg, 50 mg, 200 mg, 500 mg and bulk. Please contact Fisher for quotes. Lenvatinib mesylate (E7080 (mesylate)) is an oral and multi-targeted inhibitor of VEGFR1-3, FGFR1-4, PDGFR, KIT, and RET, with potent antitumor activities. Purity 99.03%