Learn More
Apexbio Technology LLC LENVATINIB-MESYLATE-25MG
SDP
Catalog No. 5000570414
Encompass_Preferred
lenvatinib Mesylate (CAS No 857890-39-2) is an oral tyrosine kinase inhibitor researched extensively in oncology Originating as a synthetic compound its mechanism of action involves targeting multiple receptor tyrosine kinases (RTKs) associated with tumor growth angiogenesis and progression These include VEGFR FGFR and PDGFR pathways In vitro lenvatinib Mesylate demonstrates activity in the nanomolar to low micromolar range indicating its potency It is widely applied in various cancer cell models and animal studies for testing therapeutic efficacy and understanding tumor biology Lenatatinib Mesylate is also used in drug screening programs to identify its effects and potential combinations with other therapies Experimental concentrations typically depend on the specific research design
Catalog No. 50-005-70414 Supplier Apexbio Technology LLC Supplier No. C637525
lenvatinib Mesylate (CAS No 857890-39-2) is an oral tyrosine kinase inhibitor researched extensively in oncology Originating as a synthetic compound its mechanism of action involves targeting multiple receptor tyrosine kinases (RTKs) associated with tumor growth angiogenesis and progression These include VEGFR FGFR and PDGFR pathways In vitro lenvatinib Mesylate demonstrates activity in the nanomolar to low micromolar range indicating its potency It is widely applied in various cancer cell models and animal studies for testing therapeutic efficacy and understanding tumor biology Lenatatinib Mesylate is also used in drug screening programs to identify its effects and potential combinations with other therapies Experimental concentrations typically depend on the specific research design