Learn More
Selleck Chemical LLC lenvatinib Mesylate 25mg 857890-39-2
SDP
Supplier:  Selleck Chemical LLC S524025MG
Encompass_Preferred
Lenvatinib Mesylate is a synthetic, orally available tyrosine kinase inhibitor that inhibits vascular endothelial growth factor receptor (VEGFR1-3), fibroblast growth factor receptor (FGFR1-4), platelet-derived growth factor receptor ? (PDGFR?), stem cell factor receptor (Kit (c-Kit)), and rearranged during transfection (RET (c-RET)). Lenvatinib Mesylate has potential antineoplastic activity. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8180