Learn More
Selleck Chemical LLC Lenvatinib Mesylate S5240-100mg
SDP
Catalog No. 503111721
Encompass_Preferred
Lenvatinib Mesylate is a synthetic orally available tyrosine kinase inhibitor that inhibits vascular endothelial growth factor receptor (VEGFR1-3) fibroblast growth factor receptor (FGFR1-4) platelet-derived growth factor receptor (PDGFR ) stem cell factor receptor (Kit (c-Kit)) and rearranged during transfection (RET (c-RET)) Lenvatinib Mesylate has potential antineoplastic activity
Catalog No. 50-311-1721 Supplier Selleck Chemical LLC Supplier No. S5240100MG
Lenvatinib Mesylate is a synthetic orally available tyrosine kinase inhibitor that inhibits vascular endothelial growth factor receptor (VEGFR1-3) fibroblast growth factor receptor (FGFR1-4) platelet-derived growth factor receptor (PDGFR ) stem cell factor receptor (Kit (c-Kit)) and rearranged during transfection (RET (c-RET)) Lenvatinib Mesylate has potential antineoplastic activity