Learn More
Medchemexpress LLC Lenvatinib mesylate | 857890-39-2 | 99.7% | 100 MG
SDP
Catalog No. 5000346501
Encompass_Preferred
Lenvatinib mesylate (E7080 mesylate) is an oral, multi-targeted tyrosine kinase inhibitor that inhibits VEGFR1-3, FGFR1-4, PDGFR, KIT, and RET, and shows potent antitumor activities.
  • Oral, multi-targeted tyrosine kinase inhibitor
  • Inhibits VEGFR1-3, FGFR1-4, PDGFR, KIT, and RET
  • Shows potent antitumor activities
  • 85+ Cited Publications
Catalog No. 50-003-46501 Supplier Medchemexpress LLC Supplier No. HY10981A50MG
Lenvatinib mesylate (E7080 mesylate) is an oral, multi-targeted tyrosine kinase inhibitor that inhibits VEGFR1-3, FGFR1-4, PDGFR, KIT, and RET, and shows potent antitumor activities.
  • Oral, multi-targeted tyrosine kinase inhibitor
  • Inhibits VEGFR1-3, FGFR1-4, PDGFR, KIT, and RET
  • Shows potent antitumor activities
  • 85+ Cited Publications