Learn More
Medchemexpress LLC HY-131707 5mg Medchemexpress, LEO 39652 CAS:1445656-91-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1317075MG
Encompass_Preferred
Medchemexpress, HY-131707 5mg LEO 39652 CAS:1445656-91-6 LEO 39652 is a dual-soft PDE4 inhibitor with IC50s of 1.2 nM, 1.2 nM, 3.0 nM and 3.8 nM for PDE4A, PDE4B, PDE4C and PDE4D, respectively. LEO 39652 also inhibits TNF-α with an IC50 value of 6.0 nM. LEO 39652 is used for topical research of Atopic dermatitis (AD) . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1295