Learn More
Aviva Systems Biology Corp Leo1 Antibody - N-terminal region (ARP52637_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52637_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Leo1. It was validated on Western Blot. . Alternate names: Gm185, MGC100232, MGC102277ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Yeast
Catalog No. 50-285-743