Learn More
Medchemexpress LLC HY-N0741 10mg Medchemexpress, Leonurine CAS:24697-74-3 Purity:>98%
SDP
Catalog No. 501933697
Encompass_Preferred
Medchemexpress, HY-N0741 10mg Leonurine CAS:24697-74-3 Leonurine is an alkaloid isolated from Herba leonuri, with anti-oxidative and anti-inflammatory. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3697 Supplier Medchemexpress LLC Supplier No. HYN074110MG
Medchemexpress, HY-N0741 10mg Leonurine CAS:24697-74-3 Leonurine is an alkaloid isolated from Herba leonuri, with anti-oxidative and anti-inflammatory. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.