Learn More
Medchemexpress LLC HY-N0741A 5mg Medchemexpress, Leonurine (hydrochloride) CAS:24735-18-0 Purity:>98%
SDP
Catalog No. 501933698
Encompass_Preferred
Medchemexpress, HY-N0741A 5mg Leonurine (hydrochloride) CAS:24735-18-0 Leonurine hydrochloride is an alkaloid isolated from Herba leonuri, with anti-oxidative and anti-inflammatory. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3698 Supplier Medchemexpress LLC Supplier No. HYN0741A5MG
Medchemexpress, HY-N0741A 5mg Leonurine (hydrochloride) CAS:24735-18-0 Leonurine hydrochloride is an alkaloid isolated from Herba leonuri, with anti-oxidative and anti-inflammatory. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.