Learn More
Medchemexpress LLC Leonurine hydrochloride | 24735-18-0 | 347.79 | 100 MG
SDP
Catalog No. 5000417130
Encompass_Preferred
Leonurine hydrochloride is an alkaloid isolated from Leonurus artemisia, exhibiting anti-oxidative and anti-inflammatory properties. This compound is intended for research use only, playing a role in various biological activities in vitro and in vivo.
  • Causes diminution in lipid accumulation and cellular cholesterol content.
  • Increases apoA-I- or HDL-mediated cholesterol efflux.
  • Significantly increases the expressions of ABCA1 and ABCG1.
  • Shows protective effects on cell viability of HepG2 and HL-7702 cells.
  • Improves cellular lipid accumulation via activating AMPK/SREBP1 pathway.
  • Inhibits the expression of iNOS, COX-2, PGE2, NO, TNF-α, and IL-6.
  • Suppresses ECM degradation in human OA chondrocytes.
  • Blocks IL-1β-induced PI3K and Akt phosphorylation.
  • Ameliorates osteoarthritis development in mouse DMM model.
Catalog No. 50-004-17130 Supplier Medchemexpress LLC Supplier No. HYN0741A100MG
Leonurine hydrochloride is an alkaloid isolated from Leonurus artemisia, exhibiting anti-oxidative and anti-inflammatory properties. This compound is intended for research use only, playing a role in various biological activities in vitro and in vivo.
  • Causes diminution in lipid accumulation and cellular cholesterol content.
  • Increases apoA-I- or HDL-mediated cholesterol efflux.
  • Significantly increases the expressions of ABCA1 and ABCG1.
  • Shows protective effects on cell viability of HepG2 and HL-7702 cells.
  • Improves cellular lipid accumulation via activating AMPK/SREBP1 pathway.
  • Inhibits the expression of iNOS, COX-2, PGE2, NO, TNF-α, and IL-6.
  • Suppresses ECM degradation in human OA chondrocytes.
  • Blocks IL-1β-induced PI3K and Akt phosphorylation.
  • Ameliorates osteoarthritis development in mouse DMM model.