Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Leonurine hydrochloride | 24735-18-0 | 99.4% | 347.79 | C14H22ClN3O5 | 10 MG
SDP

Catalog No. 5000296610
Encompass_Preferred

Leonurine hydrochloride is the hydrochloride salt of leonurine, an alkaloid isolated from Leonurus artemisia. Supplied as a research-grade analytical standard, it is used in biochemical and pharmacological studies exploring antioxidant and anti-inflammatory activity.

  • High purity suitable for analytical and research applications.
  • Available in small quantities for assay development and validation.
  • Stable as a sealed powder when stored at 4°C, away from moisture.
  • Molecular weight 347.79 g/mol and formula C14H22ClN3O5.
  • CAS number 24735-18-0; also known as SCM-198 hydrochloride.

Catalog No. 50-002-96610 Supplier Medchemexpress LLC Supplier No. HYN0741A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Leonurine hydrochloride is the hydrochloride salt of leonurine, an alkaloid isolated from Leonurus artemisia. Supplied as a research-grade analytical standard, it is used in biochemical and pharmacological studies exploring antioxidant and anti-inflammatory activity.

  • High purity suitable for analytical and research applications.
  • Available in small quantities for assay development and validation.
  • Stable as a sealed powder when stored at 4°C, away from moisture.
  • Molecular weight 347.79 g/mol and formula C14H22ClN3O5.
  • CAS number 24735-18-0; also known as SCM-198 hydrochloride.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.