Learn More
TARGETMOL CHEMICALS INC LEONURINE HYDROCHLORIDE 25MG
SDP
Catalog No. 502576814
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Leonurine hydrochloride (SCM-198 hydrochloride) a major alkaloid compound extracted from Leonurus japonicas Houtt. (Labiatae) is considered to have antitumor roles. purity: 99%
Catalog No. 50-257-6814 Supplier TARGETMOL CHEMICALS INC Supplier No. T408025MG
Also available in 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Leonurine hydrochloride (SCM-198 hydrochloride) a major alkaloid compound extracted from Leonurus japonicas Houtt. (Labiatae) is considered to have antitumor roles. purity: 99%