Learn More
Apexbio Technology LLC LEP (116-130) (mouse) 258276-95-8 5mg
SDP
Catalog No. 5000093299
Encompass_Preferred
LEP 116-130 (mouse) is an amidated peptide fragment derived from mouse leptin It is designed to mimic leptin-like biological activities thereby contributing to the regulation of energy balance and appetite through hypothalamic signaling pathways LEP 116-130 (mouse) exerts its biological activity by engaging pathways associated with body weight homeostasis and food consumption Based on these pharmacological properties LEP 116-130 (mouse) holds research potential in the fields of obesity metabolism regulation and related signaling pathologies
Catalog No. 50-000-93299 Supplier Apexbio Technology LLC Supplier No. A10245
LEP 116-130 (mouse) is an amidated peptide fragment derived from mouse leptin It is designed to mimic leptin-like biological activities thereby contributing to the regulation of energy balance and appetite through hypothalamic signaling pathways LEP 116-130 (mouse) exerts its biological activity by engaging pathways associated with body weight homeostasis and food consumption Based on these pharmacological properties LEP 116-130 (mouse) holds research potential in the fields of obesity metabolism regulation and related signaling pathologies