Learn More
FUJIFILM Biosciences Lepimectin Standard
Catalog No. 5000450359
Encompass_Preferred
Lepimectin is an insecticide with a 16-membered ring macrolide and considered to act on chlorine ion channels of the nerve system in insects This is an agricultural chemical reference standard for testing residual agricultural chemicals This is the mixture of Lepimectin A3 and Lepimectin A4
Catalog No. 50-004-50359 Supplier FUJIFILM Biosciences Supplier No. 12506421
Lepimectin is an insecticide with a 16-membered ring macrolide and considered to act on chlorine ion channels of the nerve system in insects This is an agricultural chemical reference standard for testing residual agricultural chemicals This is the mixture of Lepimectin A3 and Lepimectin A4