Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Leptin (93-105), human | 200436-43-7 | 98.6% | 1527.7 | 5 MG
SDP

Catalog No. 5000423384
Encompass_Preferred

Leptin (93-105), human, is a fragment of human leptin consisting of amino acids 93 to 105. Leptin is a 167-residue peptide hormone primarily produced by adipocytes, playing a crucial role in coordinating metabolic adaptations to fasting within the central nervous system. This product is for research use only.

  • White to off-white solid appearance
  • Sequence: Asn-Val-Ile-Gln-Ile-Ser-Asn-Asp-Leu-Glu-Asn-Leu-Arg (NVIQISNDLENLR)
  • Recommended storage for powder: -80°C for 2 years or -20°C for 1 year, sealed under nitrogen, away from moisture and light
  • Recommended storage for solutions: -80°C for 6 months or -20°C for 1 month, sealed under nitrogen, away from moisture and light

Catalog No. 50-004-23384 Supplier Medchemexpress LLC Supplier No. HYP25405MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Leptin (93-105), human, is a fragment of human leptin consisting of amino acids 93 to 105. Leptin is a 167-residue peptide hormone primarily produced by adipocytes, playing a crucial role in coordinating metabolic adaptations to fasting within the central nervous system. This product is for research use only.

  • White to off-white solid appearance
  • Sequence: Asn-Val-Ile-Gln-Ile-Ser-Asn-Asp-Leu-Glu-Asn-Leu-Arg (NVIQISNDLENLR)
  • Recommended storage for powder: -80°C for 2 years or -20°C for 1 year, sealed under nitrogen, away from moisture and light
  • Recommended storage for solutions: -80°C for 6 months or -20°C for 1 month, sealed under nitrogen, away from moisture and light

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.