Learn More
CPC Scientific H-Ser-Cys-Ser-Leu-Pro-Gln-Thr-Ser-Gly-Leu-Gln-Lys-Pro-Glu-Ser-NH2 (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific LEPT005B
Encompass
Peptide that is capable of mimicking the effects of recombinant leptin on body weight and food intake in female ob/ob mice that lack endogenously circulating active leptin. It also may cause body weight loss and food intake. ONE-LETTER SEQUENCE: SCSLPQTSGLQKPES-NH2MOLECULAR FORMULA: C64H109N19O24S1MOLECULAR WEIGHT:1560.75STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [258276-95-8], SYNONYMS: Leptin (116-130) amide (mouse)RESEARCH AREA: ObesityREFERENCES: P. Grasso et al., Endocrinology 138, 1413 (1997)
Catalog No. 50-210-9640