Learn More
Enzo Life Sciences Leptin (human), ELISA kit (1x96wells)
SDP
Supplier:  Enzo Life Sciences ADI900028A
Encompass_Preferred
The Leptin (human), EIA kit is a colorimetric immunometric enzyme immunoassay kit with results in 3 hours. Alternative name: OB gene product. Sensitivity: 23.4 pg/ml (range 31.3 - 2,000 pg/ml). UniProt ID: P41159. Assay Time: 3 hours. Applications: ELISA, Colorimetric detection . Species reactivity: Human . Content: Microtiter plate, Standard, Antibody, Conjugate, Assay buffer 17, TMB Substrate, Stop solution 2, Wash buffer concentrate.
Catalog No. 50-200-6828