Learn More
Enzo Life Sciences Leptin (human), (recombinant) (1mg). CAS: 177404-21-6
SDP
Supplier:  Enzo Life Sciences ALX201034M001
Encompass_Preferred
Alternative name: OB gene product. Endotoxin: <0.1EU/µg purified protein. Purity: ≥95% (SDS-PAGE; reducing and non-reducing). MW: ~16kDa. Formulation: Lyophilized from 0.0045mM sodium bicarbonate. Source: Produced in E. coli. Homologous to human serum leptin. Long Term Storage: -20°C. Use/Stability: Reconstituted protein is stable for 4 weeks when stored at +4°C. Handling: Avoid freeze/thaw cycles. After reconstitution, prepare aliquots and freeze in liquid nitrogen.
Catalog No. 50-200-7889