Learn More
Enzo Life Sciences Leptin (human), (recombinant) (>95%) (5 mg). CAS: 177404-21-6
SDP
Supplier:  Enzo Life Sciences BMLSE1610005
Encompass_Preferred
Alternative name: OB gene product. Purity: ≥95% (HPLC & SDS-PAGE). Formulation: Lyophilized. Source: Produced in E. coli. Long Term Storage: -20°C.
Catalog No. 50-201-1945