Learn More
Enzo Life Sciences Leptin (mouse), (recombinant) (1mg). CAS: 181030-10-4
SDP
Supplier:  Enzo Life Sciences ALX201035M001
Encompass_Preferred
Alternative name: OB gene product. Endotoxin: <0.1EU/µg purified protein. Purity: ≥98% (SDS-PAGE). MW: ~16kDa. Formulation: Lyophilized from 0.0045mM sodium bicarbonate. Source: Produced in E. coli. Homologous to mouse serum leptin. Long Term Storage: -20°C.
Catalog No. 50-200-7890