Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents QP53501MG
Encompass_Preferred
Product Type: Recombinant Protein. Conjugate/Tag: Untagged. Host: E. coli. Species Reactivity: Human. BioActivity: Bioactivity of Human Leptin was examined and confirmed in a Ba/F3 cell proliferation assay. This product has not been tested for in vivo use. AA Sequence: MVPIQKVQDD TKTLIKTIVT RINDISHTQS VSSKQKVTGL DFIPGLHPIL TLSKMDQTLA VYQQILTSMP SRNVIQISND LENLRDLLHV LAFSKSCHLP WASGLETLDS LGGVLEASGY STEVVALSRL QGSLQDMLWQ LDLSPGC. Buffer: Sterile filtered (0.22 um) solution containing 0.1% TFA. Purity: Greater than 95% as determined by SDS-PAGE. Endotoxin: Less than or equal to 1.00 EUs/ug as determined by the LAL method. Gene Symbol: LEP. Uniprot: Q6NT58.
Catalog No. 50-188-5786