Learn More
Enzo Life Sciences Leptin (rat), ELISA kit (1x96wells)
SDP
Supplier:  Enzo Life Sciences ADI900015A
Encompass_Preferred
The Leptin (rat), EIA kit is a colorimetric immunometric enzyme immunoassay kit with results in 3 hours. Alternative name: OB gene product. Sensitivity: 67.2 pg/ml (range 100 - 6,400 pg/ml). UniProt ID: P50596. Assay Time: 3 hours. Applications: ELISA, Colorimetric detection . Species reactivity: Rat . Content: Microtiter plate, Standard, Antibody, Conjugate, Assay buffer 28, TMB Substrate, Stop solution 2, Wash buffer concentrate.
Catalog No. 50-200-6827