Learn More
AdipoGen Leptin rat pAb Biotin
SDP
Supplier:  AdipoGen AG25A0009BC050
Encompass_Preferred
Polyclonal Antibody. Recognizes rat leptin. Weakly cross-reacts with human and mouse leptin. Detects a band of ~16kDa by Western blot. Source Rabbit. Applications ELISA, WB. Liquid. 0.2µm-filtered solution in PBS, pH 7.4. Contains no preservatives. Leptin is involved in the signaling pathway that regulates the size of the body fat depot. An increase in the level of leptin may act directly or indirectly on the CNS to inhibit food intake and/or regulate energy expenditure as part of a homeostatic mechanism to maintain constancy of the adipose mass. Defects in the leptin gene may be a cause of autosomal recessive obesity.
Catalog No. 50-256-3303