Learn More
Enzo Life Sciences Leptin receptor polyclonal antibody (100µl)
SDP
Supplier:  Enzo Life Sciences ALX210168R100
Encompass_Preferred
Host: Rabbit. Immunogen: Synthetic peptide corresponding to aa 577-594 (K577EIQWKTHEVFDAKSKSA594) of the extracellular domain of rat leptin receptor. Species reactivity: Human, Mouse, Rat. Applications: WB. Purity: Purified IgG. Formulation: Liquid. In PBS containing 0.05% sodium azide.
Catalog No. 50-200-8082