Learn More
Drg International Inc DRG Leptin Sandwich ELISA RUO, 96 Well Kit
Supplier:  Drg International Inc EIA2395R
Encompass_Preferred
An enzyme immunoassay for the quantitative measurement of leptin in serum and plasma. This product is for Research Use Only. The DRG Leptin ELISA Kit is a solid phase enzyme-linked immunosorbent assay (ELISA) based on the sandwich principle. The microtiter wells are coated with a monoclonal antibody directed towards a unique antigenic site on a Leptin molecule.
Catalog No. 50-187-1251