Learn More
ACROBiosystems Recombinant Protein;Human Leptin R / CD295 Protein, Fc Tag;HEK293;250UG;LER,LEPR,CD295,DKFZp686B1731,LEP-R,OB-R,OBR,Leptin receptor
Catalog No. 501657696
Encompass_Preferred
Recombinant Protein;Human Leptin R / CD295 Protein, Fc Tag;250UG;Human Leptin R, Fc Tag (LER-H5252) is expressed from human 293 cells (HEK293). It contains AA Phe 22 - Asp 839 (Accession # NP_002294.2).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 121 kDa. The protein migrates as 150-165 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-7696 Supplier ACROBiosystems Supplier No. LERH5252250UG
Recombinant Protein;Human Leptin R / CD295 Protein, Fc Tag;250UG;Human Leptin R, Fc Tag (LER-H5252) is expressed from human 293 cells (HEK293). It contains AA Phe 22 - Asp 839 (Accession # NP_002294.2).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 121 kDa. The protein migrates as 150-165 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.