Learn More
Cayman Chemical LeptomycIn A 100ug
SDP
Catalog No. 502985721
Encompass_Preferred
An inhibitor of nuclear transport; blocks CRM1 interaction with nuclear export signals; inhibits Rev translocation (IC50 = 0.8 nM after 7 hours); antifungal agent
Catalog No. 50-298-5721 Supplier Cayman Chemical Supplier No. 21762100 UG
An inhibitor of nuclear transport; blocks CRM1 interaction with nuclear export signals; inhibits Rev translocation (IC50 = 0.8 nM after 7 hours); antifungal agent