Learn More
Selleck Chemical LLC Leptomycin B-1mg
SDP
Supplier:  Selleck Chemical LLC S75801MG
Encompass_Preferred
Leptomycin B (LMB, CI 940, Elactocin, Mantuamycin, NSC 364372) is a potent antifungal antibiotic isolated from Streptomyces and acts as a specific inhibitor of the nuclear export factor CRM1. Leptomycin B rapidly induces cytotoxic effects in cancer cell lines via covalent inhibition of CRM1 with IC50 values of 0.1 nM–10 nM.
Catalog No. 50-238-9715