Learn More
Cayman Chemical Leptomycin B, 87081-35-4, 5 g
SDP
Supplier:  Cayman Chemical 100049765
Encompass
Molecular Formula C33H48O6, Purity ≥98%, Formula Weight 540.7, Synonyms: Elactocin, LMB, Mantuamycin, NSC 364372; A potent anti-fungal antibiotic that can directly block nuclear transport by inhibiting the action of CRM1.
Catalog No. NC1265489