Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Leptomycin B, 98%, 1mM soln. in ethanol, Thermo Scientific Chemicals

Catalog No. AAJ63784EXG
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ63784EXG 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ63784EXG Supplier Thermo Scientific Chemicals Supplier No. J63784EXG
Add to Cart

missing translation for 'validMainframeMsg'

 

Potent, specific inhibitor of nuclear export signal (NES)-dependent protein export from the nucleus

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Potent, specific inhibitor of nuclear export signal (NES)-dependent protein export from the nucleus
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 87081-35-4
Molecular Formula C33H48O6
Molecular Weight (g/mol) 540.741
MDL Number MFCD06795848
InChI Key YACHGFWEQXFSBS-BVPMNVLFSA-N
PubChem CID 121232734
IUPAC Name (2E,5S,6R,7S,9R,10E,12E,15R,16Z,18E)-17-ethyl-6-hydroxy-3,5,7,9,11,15-hexamethyl-19-[(2R,3R)-3-methyl-6-oxo-2,3-dihydropyran-2-yl]-8-oxononadeca-2,10,12,16,18-pentaenoic acid
SMILES CCC(=CC(C)CC=CC(=CC(C)C(=O)C(C)C(C(C)CC(=CC(=O)O)C)O)C)C=CC1C(C=CC(=O)O1)C

Specifications

Type Leptomycin B
Physical Form Liquid
Odor Alcohol-like
Color Colorless
Concentration 1mM soln. in ethanol
Quantity 100 μg
Flash Point 11°C (52°F)
UN Number UN1993
Merck Index 14,5444
Formula Weight 540.73
Percent Purity ≥99%
Chemical Name or Material Leptomycin B
Show More Show Less
Hazard Category H225-H319
Hazard Statement GHS H Statement
H225
Highly flammable liquid and vapor.
Precautionary Statement P210-P233-P240-P241-P242-P243-P264b-P280-P303+P361+P353-P305+P351+P338-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packaging Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C

For Manufacturing and Laboratory Use. Not for beverage use

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.