Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Leptospermone 5mg | 567-75-9 | 266.33 g/mol | C15H22O4 | 5 MG
SDP

Catalog No. 5000226888
Encompass_Preferred

Leptospermone is a natural phytochemical and a competitive, reversible inhibitor of p-hydroxyphenylpyruvate dioxygenase (HPPD). It is used as a research reagent for HPPD inhibition studies and for investigating herbicidal bleaching activity.

  • High purity (98.46%).
  • Molecular weight 266.33 g/mol.
  • Chemical formula C15H22O4.
  • Reported IC50 for HPPD inhibition: 12.1 μM.
  • Sold in small research pack sizes suitable for laboratory assays (5 mg, 10 mg, 25 mg, 50 mg).

Catalog No. 50-002-26888 Supplier Medchemexpress LLC Supplier No. HY1149915MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Leptospermone is a natural phytochemical and a competitive, reversible inhibitor of p-hydroxyphenylpyruvate dioxygenase (HPPD). It is used as a research reagent for HPPD inhibition studies and for investigating herbicidal bleaching activity.

  • High purity (98.46%).
  • Molecular weight 266.33 g/mol.
  • Chemical formula C15H22O4.
  • Reported IC50 for HPPD inhibition: 12.1 μM.
  • Sold in small research pack sizes suitable for laboratory assays (5 mg, 10 mg, 25 mg, 50 mg).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.