Learn More
ATCC Leptospira interrogans; serovar Copenhageni; genomic DNA
Catalog No. 502382974
Encompass
Genomic DNA isolated from Leptospira interrogans serovar Copenhageni strain Fiocruz L1-130 (ATCC BAA-1198). This whole-genome sequenced product can be used in PCR and other molecular biology applications.
Catalog No. 50-238-2974 Supplier ATCC Supplier No. BAA1198D5
Genomic DNA isolated from Leptospira interrogans serovar Copenhageni strain Fiocruz L1-130 (ATCC BAA-1198). This whole-genome sequenced product can be used in PCR and other molecular biology applications.