Learn More
ATCC Helicobacter pylori
Supplier:  ATCC 43629
Encompass
Helicobacter pylori strain NCTC 11639 was isolated from a gastric biopsy. This slow-growing bacterium is propagated in a microaerophilic atmosphere and requires moist conditions for best growth. Explore the use of this strain in your enteric disease research.
Catalog No. 50-238-0322