Learn More
ATCC Flavobacterium columnare
Supplier:  ATCC 23463
Encompass
Flavobacterium columnare strain NCIMB 2248 is a bacterial type strain that was isolated from diseased chinook salmon from the Snake River in Washington.
Catalog No. 50-237-9798