Learn More
Medchemexpress LLC Lercanidipine-d3 hydrochloride | 1189954-18-4 | 98.14% | 1 MG
SDP
Catalog No. 5000407728
Encompass_Preferred
Lercanidipine-d3 hydrochloride is the deuterium-labeled form of Lercanidipine, a lipophilic third-generation dihydropyridine-calcium channel blocker (DHP-CCB). It offers long-lasting antihypertensive action and reno-protective effects. Stable heavy isotopes of hydrogen, carbon, and other elements are incorporated into drug molecules primarily as tracers for quantitation during drug development. Deuteration can impact the pharmacokinetic and metabolic profiles of drugs.
  • Functions as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-07728 Supplier Medchemexpress LLC Supplier No. HYB0612DS11MG
Lercanidipine-d3 hydrochloride is the deuterium-labeled form of Lercanidipine, a lipophilic third-generation dihydropyridine-calcium channel blocker (DHP-CCB). It offers long-lasting antihypertensive action and reno-protective effects. Stable heavy isotopes of hydrogen, carbon, and other elements are incorporated into drug molecules primarily as tracers for quantitation during drug development. Deuteration can impact the pharmacokinetic and metabolic profiles of drugs.
  • Functions as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS