Learn More
Medchemexpress LLC Lercanidipine hydrochloride | 132866-11-6 | 1 ML
SDP
Catalog No. 5000407727
Encompass_Preferred
Lercanidipine hydrochloride is a third-generation, lipophilic, brain-penetrant, vascular-selective dihydropyridine-calcium channel blocker. It exhibits long-lasting antihypertensive action, reno- and neuro-protective effects, and anti-oxidant, anti-inflammatory, and anti-apoptotic properties. It is suitable for cardiovascular and neurological research.
  • Calcium channel blocker
  • Antihypertensive action
  • Reno- and neuro-protective effects
  • Anti-oxidant properties
  • Anti-inflammatory properties
  • Anti-apoptotic properties
  • Suitable for cardiovascular and neurological research
Catalog No. 50-004-07727 Supplier Medchemexpress LLC Supplier No. HYB0612A10MM/1ML
Lercanidipine hydrochloride is a third-generation, lipophilic, brain-penetrant, vascular-selective dihydropyridine-calcium channel blocker. It exhibits long-lasting antihypertensive action, reno- and neuro-protective effects, and anti-oxidant, anti-inflammatory, and anti-apoptotic properties. It is suitable for cardiovascular and neurological research.
  • Calcium channel blocker
  • Antihypertensive action
  • Reno- and neuro-protective effects
  • Anti-oxidant properties
  • Anti-inflammatory properties
  • Anti-apoptotic properties
  • Suitable for cardiovascular and neurological research