Learn More
Apexbio Technology LLC Lercanidipine hydrochloride 132866-11-6 200mg
SDP
Supplier:  Apexbio Technology LLC B3482200
Encompass_Preferred
Lercanidipine hydrochloride (CAS 132866-11-6) is a dihydropyridine calcium channel blocker that selectively inhibits L-type calcium channels reducing intracellular calcium influx and leading to vasodilation and decreased blood pressure Due to its lipophilic nature it can cross the blood-brain barrier exhibiting neuroprotective and renoprotective properties In addition to vascular effects lercanidipine hydrochloride has demonstrated antioxidative anti-inflammatory and anti-apoptotic activities in preclinical studies It is widely utilized in research on cardiovascular regulation neuroprotection inflammatory processes and vascular dysfunction
Catalog No. 50-000-94497