Learn More
Medchemexpress LLC Lercanidipine (hydrochloride) | 132866-11-6 | 99.93% | 250 MG
SDP
Catalog No. 5000406528
Encompass_Preferred
Lercanidipine hydrochloride is a third-generation, lipophilic, brain-penetrant, vascular-selective, and orally active dihydropyridine-calcium channel blocker with a pIC50 of 7.74. It exhibits long-lasting antihypertensive action and offers reno- and neuro-protective effects, and can be used in cardiovascular and neurological research.
  • Functions as a dihydropyridine-calcium channel blocker
  • Demonstrates long-lasting antihypertensive action
  • Provides reno- and neuro-protective effects
  • Exhibits anti-oxidant properties
  • Possesses anti-inflammatory properties
  • Shows anti-apoptotic properties
  • Suitable for cardiovascular and neurological research
Catalog No. 50-004-06528 Supplier Medchemexpress LLC Supplier No. 250MGHYB0612A
Lercanidipine hydrochloride is a third-generation, lipophilic, brain-penetrant, vascular-selective, and orally active dihydropyridine-calcium channel blocker with a pIC50 of 7.74. It exhibits long-lasting antihypertensive action and offers reno- and neuro-protective effects, and can be used in cardiovascular and neurological research.
  • Functions as a dihydropyridine-calcium channel blocker
  • Demonstrates long-lasting antihypertensive action
  • Provides reno- and neuro-protective effects
  • Exhibits anti-oxidant properties
  • Possesses anti-inflammatory properties
  • Shows anti-apoptotic properties
  • Suitable for cardiovascular and neurological research