Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Lercanidipine hydrochloride, 98%

Catalog No. AAH3672803
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH36728-03 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH36728-03 Supplier Thermo Scientific™ Supplier No. H3672803
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

CAS 132866-11-6
Melting Point 175-177°
Molecular Formula C36H42ClN3O6
Quantity 1 g
MDL Number MFCD07773089
UN Number UN2811
InChI Key WMFYOYKPJLRMJI-UHFFFAOYNA-N
SMILES [H+].[Cl-].COC(=O)C1=C(C)NC(C)=C(C1C1=CC=CC(=C1)[N+]([O-])=O)C(=O)OC(C)(C)CN(C)CCC(C1=CC=CC=C1)C1=CC=CC=C1
IUPAC Name hydrogen 3-{1-[(3,3-diphenylpropyl)(methyl)amino]-2-methylpropan-2-yl} 5-methyl 2,6-dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate chloride
Molecular Weight (g/mol) 648.20
Formula Weight 648.19
Percent Purity 98%
Physical Form Solid
Show More Show Less
ShelfLife 5 Years
EINECSNumber 603-682-3
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.