Learn More
Cayman Chemical Lercanidipine (hydrochloride), Purity: ≥98%, CAS: 132866-11-6, 100 mg
SDP
Catalog No. 502941913
Encompass_Preferred
A dihydropyridine L-type calcium channel blocker; binds to rat brain and heart homogenates (Kis = 0.24-0.3 and 0.22 nM, respectively, in radioligand binding assays) and inhibits potassium-induced contraction of isolated rat aorta (IC50 = 1.3 nM); reduces DBP in normotensive and spontaneously hypertensive rats (ED25s = 16.3 and 15.5 µg/kg, respectively)
Catalog No. 50-294-1913 Supplier Cayman Chemical Supplier No. 29104100 MG
A dihydropyridine L-type calcium channel blocker; binds to rat brain and heart homogenates (Kis = 0.24-0.3 and 0.22 nM, respectively, in radioligand binding assays) and inhibits potassium-induced contraction of isolated rat aorta (IC50 = 1.3 nM); reduces DBP in normotensive and spontaneously hypertensive rats (ED25s = 16.3 and 15.5 µg/kg, respectively)