Learn More
Medchemexpress LLC Leriglitazone (hydrochloride) | 146062-46-6 | 408.90 | C19H21ClN2O4S | 10 MG
SDP
Catalog No. 5000228784
Encompass_Preferred
Leriglitazone hydrochloride is the hydrochloride salt of leriglitazone (MIN-102), a blood-brain barrier-penetrant PPARγ agonist used in preclinical research to study neuroinflammation and neurodegeneration. It modulates mitochondrial function and shows neuroprotective, anti-inflammatory, and antioxidant effects.
  • Hydrochloride salt suitable for research use.
  • PPARγ agonist with reported EC50 ~9 μM.
  • Blood-brain barrier penetrant and orally active in reported studies.
  • Used to study neuroinflammatory and neurodegenerative disease mechanisms.
  • Molecular weight approximately 408.90 and formula C19H21ClN2O4S (from SDS).
Catalog No. 50-002-28784 Supplier Medchemexpress LLC Supplier No. HY117727A10MG
Leriglitazone hydrochloride is the hydrochloride salt of leriglitazone (MIN-102), a blood-brain barrier-penetrant PPARγ agonist used in preclinical research to study neuroinflammation and neurodegeneration. It modulates mitochondrial function and shows neuroprotective, anti-inflammatory, and antioxidant effects.
  • Hydrochloride salt suitable for research use.
  • PPARγ agonist with reported EC50 ~9 μM.
  • Blood-brain barrier penetrant and orally active in reported studies.
  • Used to study neuroinflammatory and neurodegenerative disease mechanisms.
  • Molecular weight approximately 408.90 and formula C19H21ClN2O4S (from SDS).