Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Leriglitazone hydrochloride | 146062-46-6 | 99.6% | 408.90 | C19H21ClN2O4S | 50 MG
SDP

Catalog No. 5000228786
Encompass_Preferred

Leriglitazone hydrochloride is a research-grade PPARγ agonist used to investigate neuroprotective, anti-inflammatory, and antioxidant mechanisms. Supplied as a solid for laboratory use, the compound is characterized for purity and recommended storage conditions.

  • High purity: 99.6%.
  • PPARγ agonist with neuroprotective and anti-inflammatory activity.
  • Solid, white to light yellow, suitable for handling in laboratory assays.
  • Available in milligram-scale packages for research studies.
  • Stable when stored sealed and protected from moisture; in solvent, -80 °C for up to six months.
  • Molecular weight 408.90; formula C19H21ClN2O4S.

Catalog No. 50-002-28786 Supplier Medchemexpress LLC Supplier No. HY117727A50MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Leriglitazone hydrochloride is a research-grade PPARγ agonist used to investigate neuroprotective, anti-inflammatory, and antioxidant mechanisms. Supplied as a solid for laboratory use, the compound is characterized for purity and recommended storage conditions.

  • High purity: 99.6%.
  • PPARγ agonist with neuroprotective and anti-inflammatory activity.
  • Solid, white to light yellow, suitable for handling in laboratory assays.
  • Available in milligram-scale packages for research studies.
  • Stable when stored sealed and protected from moisture; in solvent, -80 °C for up to six months.
  • Molecular weight 408.90; formula C19H21ClN2O4S.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.